Anti-TRIM49 Mouse Monoclonal Antibody [clone: 200000]

Supplier: Abnova
H00057093-M01
10604-682EA 502.5 CAD
10604-682
Anti-TRIM49 Mouse Monoclonal Antibody [clone: 200000]
Antibodies
Type: Primary
Antigen: TRIM49
Clonality: Monoclonal
Clone: 200000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR