Anti-TRIM49 Mouse Monoclonal Antibody [clone: 3C7]

Supplier: Abnova
H00057093-M05
10604-680EA 502.5 CAD
10604-680
Anti-TRIM49 Mouse Monoclonal Antibody [clone: 3C7]
Antibodies
Type: Primary
Antigen: TRIM49
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR