Anti-CD177 Mouse Polyclonal Antibody

H00057126-B01P
10560-186EA 420.99 CAD
10560-186
Anti-CD177 Mouse Polyclonal Antibody
Type: Primary
Antigen: CD177
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR