Anti-SCYL3 Mouse Monoclonal Antibody [clone: 3D3]

Supplier: Abnova
H00057147-M01
10600-786EA 502.5 CAD
10600-786
Anti-SCYL3 Mouse Monoclonal Antibody [clone: 3D3]
Antibodies
Type: Primary
Antigen: SCYL3
Clonality: Monoclonal
Clone: 3D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR