Anti-MRS2 Mouse Monoclonal Antibody [clone: 100]

Supplier: Abnova
H00057380-M03
10593-838EA 502.5 CAD
10593-838
Anti-MRS2 Mouse Monoclonal Antibody [clone: 100]
Antibodies
Type: Primary
Antigen: MRS2
Clonality: Monoclonal
Clone: 100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR