Anti-CLK4 Mouse Polyclonal Antibody

H00057396-B01P
10553-662EA 420.99 CAD
10553-662
Anti-CLK4 Mouse Polyclonal Antibody
Type: Primary
Antigen: CLK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR