Anti-SPC25 Mouse Polyclonal Antibody

H00057405-B01P
10579-414EA 420.99 CAD
10579-414
Anti-SPC25 Mouse Polyclonal Antibody
Type: Primary
Antigen: SPC25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR