Anti-ESYT2 Mouse Monoclonal Antibody [clone: 4C3]

Supplier: Abnova
H00057488-M02
10591-046EA 502.5 CAD
10591-046
Anti-ESYT2 Mouse Monoclonal Antibody [clone: 4C3]
Antibodies
Type: Primary
Antigen: ESYT2
Clonality: Monoclonal
Clone: 4C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR