Anti-ARID1B Mouse Monoclonal Antibody [clone: 2D2]

Supplier: Abnova
H00057492-M01
10577-264EA 502.5 CAD
10577-264
Anti-ARID1B Mouse Monoclonal Antibody [clone: 2D2]
Antibodies
Type: Primary
Antigen: ARID1B
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR