Anti-XPO5 Mouse Monoclonal Antibody [clone: 2C5-1B3]

Supplier: Abnova
H00057510-M01
10574-036EA 502.5 CAD
10574-036
Anti-XPO5 Mouse Monoclonal Antibody [clone: 2C5-1B3]
Antibodies
Type: Primary
Antigen: XPO5
Clonality: Monoclonal
Clone: 2C5-1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR