Anti-RPTOR Mouse Monoclonal Antibody [clone: 3C2]

Supplier: Abnova
H00057521-M04
10600-402EA 502.5 CAD
10600-402
Anti-RPTOR Mouse Monoclonal Antibody [clone: 3C2]
Antibodies
Type: Primary
Antigen: RPTOR
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR