Anti-SRGAP1 Mouse Monoclonal Antibody [clone: 5D10]

Supplier: Abnova
H00057522-M07
10602-072EA 502.5 CAD
10602-072
Anti-SRGAP1 Mouse Monoclonal Antibody [clone: 5D10]
Antibodies
Type: Primary
Antigen: SRGAP1
Clonality: Monoclonal
Clone: 5D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR