Anti-TAOK1 Mouse Monoclonal Antibody [clone: 4E12]

Supplier: Abnova
H00057551-M01
10603-244EA 502.5 CAD
10603-244
Anti-TAOK1 Mouse Monoclonal Antibody [clone: 4E12]
Antibodies
Type: Primary
Antigen: TAOK1
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR