Anti-SEMA6A Mouse Polyclonal Antibody

H00057556-B01
10570-854EA 392.79 CAD
10570-854
Anti-SEMA6A Mouse Polyclonal Antibody
Type: Primary
Antigen: SEMA6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR