Anti-VPS18 Mouse Monoclonal Antibody [clone: 2G10]

Supplier: Abnova
H00057617-M02A
10625-590EA 502.5 CAD
10625-590
Anti-VPS18 Mouse Monoclonal Antibody [clone: 2G10]
Antibodies
Type: Primary
Antigen: VPS18
Clonality: Monoclonal
Clone: 2G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR