Anti-SEMA4G Mouse Monoclonal Antibody [clone: 3F2]

Supplier: Abnova
H00057715-M01
10600-904EA 502.5 CAD
10600-904
Anti-SEMA4G Mouse Monoclonal Antibody [clone: 3F2]
Antibodies
Type: Primary
Antigen: SEMA4G
Clonality: Monoclonal
Clone: 3F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR