Anti-PCDHB16 Mouse Monoclonal Antibody [clone: 3F12-2A6]

Supplier: Abnova
H00057717-M01
10596-248EA 502.5 CAD
10596-248
Anti-PCDHB16 Mouse Monoclonal Antibody [clone: 3F12-2A6]
Antibodies
Type: Primary
Antigen: PCDHB16
Clonality: Monoclonal
Clone: 3F12-2A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR