Anti-METTL14 Mouse Polyclonal Antibody

H00057721-B01P
10562-392EA 420.99 CAD
10562-392
Anti-METTL14 Mouse Polyclonal Antibody
Type: Primary
Antigen: METTL14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR