Anti-IGDCC4 Mouse Monoclonal Antibody [clone: 6A3]

Supplier: Abnova
H00057722-M05
10595-124EA 502.5 CAD
10595-124
Anti-IGDCC4 Mouse Monoclonal Antibody [clone: 6A3]
Antibodies
Type: Primary
Antigen: IGDCC4
Clonality: Monoclonal
Clone: 6A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR