Anti-WDR19 Rabbit Polyclonal Antibody

Supplier: Abnova
H00057728-D01
10605-754EA 564 CAD
10605-754
Anti-WDR19 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: WDR19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR