Anti-GBA3 Mouse Polyclonal Antibody

Supplier: Abnova
H00057733-A01
10557-378EA 364.5 CAD
10557-378
Anti-GBA3 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GBA3 (glucosidase, beta, acid 3 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR