Anti-CCAR2 Rabbit Polyclonal Antibody

Supplier: Abnova
H00057805-D01P
10590-462EA 564 CAD
10590-462
Anti-CCAR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR