Anti-CXCL16 Mouse Monoclonal Antibody [clone: 2B12]

Supplier: Abnova
H00058191-M05
10583-284EA 502.5 CAD
10583-284
Anti-CXCL16 Mouse Monoclonal Antibody [clone: 2B12]
Antibodies
Type: Primary
Antigen: CXCL16
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR