Anti-PCTP Mouse Monoclonal Antibody [clone: 1F9]

Supplier: Abnova
H00058488-M01A
10624-892EA 502.5 CAD
10624-892
Anti-PCTP Mouse Monoclonal Antibody [clone: 1F9]
Antibodies
Type: Primary
Antigen: PCTP
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR