Anti-RPRD1B Mouse Monoclonal Antibody [clone: 4D6]

Supplier: Abnova
H00058490-M04
10578-314EA 502.5 CAD
10578-314
Anti-RPRD1B Mouse Monoclonal Antibody [clone: 4D6]
Antibodies
Type: Primary
Antigen: RPRD1B
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR