Anti-IL21 Mouse Polyclonal Antibody

H00059067-B01P
10561-668EA 420.99 CAD
10561-668
Anti-IL21 Mouse Polyclonal Antibody
Type: Primary
Antigen: IL21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR