Anti-SIGIRR Rabbit Polyclonal Antibody

Supplier: Abnova
H00059307-D01
10601-400EA 564 CAD
10601-400
Anti-SIGIRR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SIGIRR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR