Anti-ZNF350 Mouse Monoclonal Antibody [clone: 1A9]

Supplier: Abnova
H00059348-M01
10606-496EA 502.5 CAD
10606-496
Anti-ZNF350 Mouse Monoclonal Antibody [clone: 1A9]
Antibodies
Type: Primary
Antigen: ZNF350
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR