Anti-SAV1 Mouse Monoclonal Antibody [clone: 3B2]

Supplier: Abnova
H00060485-M02
10600-668EA 502.5 CAD
10600-668
Anti-SAV1 Mouse Monoclonal Antibody [clone: 3B2]
Antibodies
Type: Primary
Antigen: SAV1
Clonality: Monoclonal
Clone: 3B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR