Anti-ELAC2 Mouse Monoclonal Antibody [clone: 1A2]

Supplier: Abnova
H00060528-M01
10584-890EA 502.5 CAD
10584-890
Anti-ELAC2 Mouse Monoclonal Antibody [clone: 1A2]
Antibodies
Type: Primary
Antigen: ELAC2
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR