Anti-CELF5 Mouse Polyclonal Antibody

Supplier: Abnova
H00060680-B01
10551-738EA 502.5 CAD
10551-738
Anti-CELF5 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CELF5
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR