Anti-FKBP10 Mouse Monoclonal Antibody [clone: 3B5]

Supplier: Abnova
H00060681-M02
10586-076EA 502.5 CAD
10586-076
Anti-FKBP10 Mouse Monoclonal Antibody [clone: 3B5]
Antibodies
Type: Primary
Antigen: FKBP10 (FK506 binding protein 10, 65 kDa)
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR