Anti-NEUROG2 Mouse Monoclonal Antibody [clone: 1G8]

Supplier: Abnova
H00063973-M03
10594-736EA 502.5 CAD
10594-736
Anti-NEUROG2 Mouse Monoclonal Antibody [clone: 1G8]
Antibodies
Type: Primary
Antigen: NEUROG2 (neurogenin 2)
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR