Anti-SAMSN1 Mouse Monoclonal Antibody [clone: 1B8]

Supplier: Abnova
H00064092-M01
10600-640EA 502.5 CAD
10600-640
Anti-SAMSN1 Mouse Monoclonal Antibody [clone: 1B8]
Antibodies
Type: Primary
Antigen: SAMSN1
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR