Anti-CENPK Mouse Monoclonal Antibody [clone: 4D11]

Supplier: Abnova
H00064105-M08
10586-106EA 502.5 CAD
10586-106
Anti-CENPK Mouse Monoclonal Antibody [clone: 4D11]
Antibodies
Type: Primary
Antigen: CENPK
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR