Anti-CRLF2 Mouse Monoclonal Antibody [clone: 4A11]

Supplier: Abnova
H00064109-M03
10582-694EA 502.5 CAD
10582-694
Anti-CRLF2 Mouse Monoclonal Antibody [clone: 4A11]
Antibodies
Type: Primary
Antigen: CRLF2
Clonality: Monoclonal
Clone: 4A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR