Anti-MOAP1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00064112-D01P
10592-970EA 564 CAD
10592-970
Anti-MOAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MOAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR