Anti-MOAP1 Mouse Monoclonal Antibody [clone: 4A11]

Supplier: Abnova
H00064112-M01
10592-966EA 502.5 CAD
10592-966
Anti-MOAP1 Mouse Monoclonal Antibody [clone: 4A11]
Antibodies
Type: Primary
Antigen: MOAP1
Clonality: Monoclonal
Clone: 4A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR