Anti-NCAPG Mouse Monoclonal Antibody [clone: 4B1]

Supplier: Abnova
H00064151-M01
10588-376EA 502.5 CAD
10588-376
Anti-NCAPG Mouse Monoclonal Antibody [clone: 4B1]
Antibodies
Type: Primary
Antigen: NCAPG
Clonality: Monoclonal
Clone: 4B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR