Anti-NECAB1 Mouse Polyclonal Antibody

H00064168-B01P
10555-634EA 420.99 CAD
10555-634
Anti-NECAB1 Mouse Polyclonal Antibody
Type: Primary
Antigen: NECAB1 (N-terminal EF-hand calcium binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR