Anti-CARD9 Mouse Polyclonal Antibody

H00064170-B01P
10552-526EA 420.99 CAD
10552-526
Anti-CARD9 Mouse Polyclonal Antibody
Type: Primary
Antigen: CARD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR