Anti-P3H1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00064175-D01P
10591-680EA 564 CAD
10591-680
Anti-P3H1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: P3H1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR