Anti-P3H1 Mouse Monoclonal Antibody [clone: 3C7]

Supplier: Abnova
H00064175-M01
10592-420EA 502.5 CAD
10592-420
Anti-P3H1 Mouse Monoclonal Antibody [clone: 3C7]
Antibodies
Type: Primary
Antigen: P3H1
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR