Anti-SEMA4A (sema Domain, immunoglobulin Domain (Ig), transmembrane Domain (TM) And short cytoplasmic Domain, (semaphorin) 4A) Mouse Polyclonal Antibody

H00064218-B01P
10570-840EA 420.99 CAD
10570-840
Anti-SEMA4A (sema Domain, immunoglobulin Domain (Ig), transmembrane Domain (TM) And short cytoplasmic Domain, (semaphorin) 4A) Mouse Polyclonal Antibody
Type: Primary
Antigen: SEMA4A (sema Domain, immunoglobulin Domain (Ig), transmembrane Domain (TM) And short cytoplasmic Domain, (semaphorin) 4A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR