Anti-STRA6 Rabbit Polyclonal Antibody

Supplier: Abnova
H00064220-D01P
10602-798EA 564 CAD
10602-798
Anti-STRA6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: STRA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR