Anti-ZSCAN31 Mouse Monoclonal Antibody [clone: 4F7]

Supplier: Abnova
H00064288-M03
10606-482EA 502.5 CAD
10606-482
Anti-ZSCAN31 Mouse Monoclonal Antibody [clone: 4F7]
Antibodies
Type: Primary
Antigen: ZSCAN31
Clonality: Monoclonal
Clone: 4F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR