Anti-RFWD2 Mouse Monoclonal Antibody [clone: 10000]

Supplier: Abnova
H00064326-M01
10599-618EA 502.5 CAD
10599-618
Anti-RFWD2 Mouse Monoclonal Antibody [clone: 10000]
Antibodies
Type: Primary
Antigen: RFWD2
Clonality: Monoclonal
Clone: 10000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR