Anti-AKTIP Mouse Monoclonal Antibody [clone: 6G8]

Supplier: Abnova
H00064400-M03
10586-806EA 502.5 CAD
10586-806
Anti-AKTIP Mouse Monoclonal Antibody [clone: 6G8]
Antibodies
Type: Primary
Antigen: AKTIP
Clonality: Monoclonal
Clone: 6G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR