Anti-VPS16 Mouse Monoclonal Antibody [clone: 2F10]

Supplier: Abnova
H00064601-M05
10605-660EA 502.5 CAD
10605-660
Anti-VPS16 Mouse Monoclonal Antibody [clone: 2F10]
Antibodies
Type: Primary
Antigen: VPS16
Clonality: Monoclonal
Clone: 2F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR