Anti-CREB3L2 Mouse Monoclonal Antibody [clone: 1B8]

Supplier: Abnova
H00064764-M03
10582-866EA 502.5 CAD
10582-866
Anti-CREB3L2 Mouse Monoclonal Antibody [clone: 1B8]
Antibodies
Type: Primary
Antigen: CREB3L2
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR